Methyl-5-bromopicolinate ≥98%
Supplier: BeanTown Chemical
Synonyms:
5-Bromopyridine-2-carboxylic acid methyl ester, Methyl 5-bromopyridine-2-carboxylate, Methyl 5-bromopicolinate
130945-1G
130945-5G
BT130945-1GEA
62
USD
BT130945-1G
BT130945-5G
Methyl-5-bromopicolinate ≥98%
Methyl 5-bromopicolinate
CAS: 29682-15-3; MDL No: MFCD04112493
Solid; Molecular Formula: C7H6BrNO2 ; MW: 216.03
Melting Point: 99-103°
- Solid
Formula:
C₇H₆BrNO₂ MW: 216.03 g/mol Melting Pt: 99…103 °C |
MDL Number:
MFCD04112493 CAS Number: 29682-15-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...