(2,4-Pentanedionato)lithium (In₂O₃:SnO₂) (90:10%) (w/w) ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Lithium acetylacetonate, Acetylacetone lithium salt

130940-10G 130940-250G 130940-50G
BT130940-10GEA 20 USD
BT130940-10G BT130940-250G BT130940-50G
(2,4-Pentanedionato)lithium (In₂O₃:SnO₂) (90:10%) (w/w) ≥99.5%
(2,4-Pentanedionato)lithium

CAS: 18115-70-3; EC No: 242-008-9; MDL No: MFCD00013496 Crystalline; Molecular Formula: C5H7LiO2 ; MW: 106.05 Melting Point: 250° (decomposes) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₅H₇LiO₂
MW: 106.05 g/mol
Melting Pt: 250 °C (dec.)
MDL Number: MFCD00013496
CAS Number: 18115-70-3
EINECS: 242-008-9

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR