Strontium acetate hydrate ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
130925-25G
130925-5G
BT130925-25GEA
302
USD
BT130925-25G
BT130925-5G
Strontium acetate hydrate ≥99.995% (trace metals basis)
Strontium acetate hydrate
CAS: 543-94-2; EC No: 208-854-8; MDL No: MFCD00036392; RTECS: AJ4725000
Crystalline; Linear Formula: (CH3COO)2Sr·xH2O; MW: 205.71 (anhydrous)
Hygroscopic
- RTECS: AJ4725000
- Hygroscopic
- Crystalline
Formula:
C₄H₆O₄Sr·xH₂O |
MDL Number:
MFCD00036392 CAS Number: 543-94-2 EINECS: 208-854-8 |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...