Molybdenum(IV) sulphide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Molybdenum disulphide
130900-100G
130900-25G
130900-500G
BT130900-100GEA
57.08
USD
BT130900-100G
BT130900-25G
BT130900-500G
Molybdenum(IV) sulphide ≥99%
Molybdenum(IV) sulphide
CAS: 1317-33-5; EC No: 215-263-9; MDL No: MFCD00003470
-325 Mesh Powder; Molecular Formula: MoS2; MW: 160.08
Melting Point: 2375°
Density (g/mL): 5.06
- -325 Mesh Powder
Formula:
MoS₂ Melting Pt: 2375 °C Density: 5.06 g/mL |
MDL Number:
MFCD00003470 CAS Number: 1317-33-5 EINECS: 215-263-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...