3-Bromo-4-methylpyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Bromo-4-picoline

130890-1G 130890-25G 130890-5G
BT130890-1GEA 37 USD
BT130890-1G BT130890-25G BT130890-5G
3-Bromo-4-methylpyridine ≥98%
3-Bromo-4-methylpyridine

CAS: 3430-22-6; MDL No: MFCD00082592 Liquid; Molecular Formula: C6H6BrN; MW: 172.02 Boiling Point: 199-200°; Flash point: 79°C (174°F) Density (g/mL): 1.549; Refractive Index: 1.56


  • Refractive index: 1.56
  • Liquid

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 199…200 °C
Density: 1.549 g/mL
Flash Pt: 79 °C (174 °F)
MDL Number: MFCD00082592
CAS Number: 3430-22-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR