3,5-Dichlorobenzyl alcohol ≥98%
Supplier: BeanTown Chemical
130850-25G
130850-5G
BT130850-25GEA
290
USD
BT130850-25G
BT130850-5G
3,5-Dichlorobenzyl alcohol ≥98%
3,5-Dichlorobenzyl alcohol
CAS: 60211-57-6; EC No: 262-109-1; MDL No: MFCD00004634
Solid; Molecular Formula: C7H6Cl2O; MW: 177.03
Melting Point: 79-82°
- Solid
Formula:
C₇H₆Cl₂O MW: 177.03 g/mol Melting Pt: 79…82 °C |
MDL Number:
MFCD00004634 CAS Number: 60211-57-6 EINECS: 262-109-1 UN: 2250 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...