5-Chlorosalicylaldehyde ≥98%
Supplier: BeanTown Chemical
130840-100G
130840-25G
130840-5G
BT130840-100GEA
379
USD
BT130840-100G
BT130840-25G
BT130840-5G
5-Chlorosalicylaldehyde ≥98%
5-Chlorosalicylaldehyde
CAS: 635-93-8; EC No: 211-244-4; MDL No: MFCD00003331; RTECS: VN5450000
Solid; Molecular Formula: C7H5ClO2 ; MW: 156.57
Melting Point: 100-102°
Air Sensitive
- RTECS: VN5450000
- Air Sensitive
- Solid
Formula:
C₇H₅ClO₂ Melting Pt: 100…102 °C |
MDL Number:
MFCD00003331 CAS Number: 635-93-8 EINECS: 211-244-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...