Aluminium stearate, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Aluminum stearate, Aluminum tristearate

130815-1KG 130815-5KG
BT130815-1KGEA 69.12 USD
BT130815-1KG BT130815-5KG
Aluminium stearate, Technical Grade
Aluminium stearate

CAS: 637-12-7; EC No: 211-279-5; MDL No: MFCD00036389 Powder; Linear Formula: Al[CH3(CH2)16CO2]3 ; Molecular Formula: C54H105AlO6; MW: 877.42 Melting Point: 105-115°


  • Powder

Formula: C₅₄H₁₀₅AlO₆
MW: 877.389 g/mol
Melting Pt: 105…115 °C
MDL Number: MFCD00036389
CAS Number: 637-12-7
EINECS: 211-279-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR