Diallyldimethylammonium chloride 65% (w/w) in water

Supplier: BeanTown Chemical
Warning

130760-1L 130760-3L
BT130760-1LEA 134.3 USD
BT130760-1L BT130760-3L
Diallyldimethylammonium chloride 65% (w/w) in water
Diallyldimethylammonium chloride

CAS: 7398-69-8; MDL No: MFCD00043200 Liquid; Linear Formula: [(CH3)2N(CH2CH=CH2)2]Cl; Molecular Formula: C8H16ClN; MW: 161.67


  • Liquid

Formula: C₈H₁₆ClN
MW: 161.67 g/mol
MDL Number: MFCD00043200
CAS Number: 7398-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR