4-Chloro-2-(trifluoromethyl)quinazoline ≥98%

Supplier: BeanTown Chemical

130715-1G 130715-250MG
BT130715-1GEA 335 USD
BT130715-1G BT130715-250MG
4-Chloro-2-(trifluoromethyl)quinazoline ≥98%
4-Chloro-2-(trifluoromethyl)quinazoline

CAS: 52353-35-2; MDL No: MFCD00793683 Powder; Molecular Formula: C9H4ClF3N2; MW: 232.59


  • Powder

Formula: C₉H₄ClF₃N₂
MW: 232.5900 g/mol
MDL Number: MFCD00793683
CAS Number: 52353-35-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR