Indium (III) oxide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Indium trioxide, Diindium trioxide

130670-10G 130670-250G 130670-50G
BT130670-10GEA 66 USD
BT130670-10G BT130670-250G BT130670-50G
Indium (III) oxide ≥99.9% (trace metals basis)
Indium (III) oxide

CAS: 1312-43-2; EC No: 215-193-9; MDL No: MFCD00011060 -325 Mesh Powder; Molecular Formula: In2O3; MW: 277.64 Density (g/mL): 7.18


  • -325 Mesh Powder

Formula: In₂O₃
MW: 277.64 g/mol
Density: 7.18 g/mL
MDL Number: MFCD00011060
CAS Number: 1312-43-2
EINECS: 215-193-9

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR