Tetrasodium diphosphate, anhydrous, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
Sodium pyrophosphate tetrabasic, Tetrasodium diphosphate, Sodium diphosphate, Sodium pyrophosphate, tetra-Sodium pyrophosphate
130655-1KG
130655-250G
BT130655-1KGEA
77.22
USD
BT130655-1KG
BT130655-250G
Tetrasodium diphosphate, anhydrous, Technical Grade
Tetrasodium diphosphate
CAS: 7722-88-5; EC No: 231-767-1; MDL No: MFCD00003513; RTECS: UX7350000
Granular; Linear Formula: Na4P2O7; MW: 265.90
Melting Point: 622-985°
Density (g/mL): 2.534
Hygroscopic
- RTECS: UX7350000
- Hygroscopic
- Granular
Formula:
Na₄O₇P₂ MW: 265.9 g/mol Melting Pt: 622…985 °C Density: 2.534 g/mL |
MDL Number:
MFCD00003513 CAS Number: 7722-88-5 EINECS: 231-767-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...