β-Chloropropiophenone ≥96%

Supplier: BeanTown Chemical

130645-10G 130645-50G
BT130645-10GEA 49 USD
BT130645-10G BT130645-50G
β-Chloropropiophenone ≥96%
β-Chloropropiophenone

CAS: 936-59-4; EC No: 213-317-6; MDL No: MFCD00000990 Crystalline; Molecular Formula: C9H9ClO; MW: 168.62 Melting Point: 48-50°; Boiling Point: 113-115°/4 mmHg; Flash point: 110°C (230°F)


  • Crystalline

Formula: (C₆H₅)C(O)CH₂CH₂Cl
Boiling Pt: 113…115 °C/4 mmHg
Melting Pt: 48…50 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00000990
CAS Number: 936-59-4
EINECS: 213-317-6

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR