Cesium sulphate ≥99.99%

Supplier: BeanTown Chemical

130620-100G 130620-25G
BT130620-100GEA 203 USD
BT130620-100G BT130620-25G
Cesium sulphate ≥99.99%
Cesium sulphate

CAS: 10294-54-9; EC No: 233-662-6; MDL No: MFCD00010959 -60 Mesh Powder; Linear Formula: Cs2SO4; MW: 361.87 Melting Point: 1019° Density (g/mL): 4.243 Hygroscopic


  • Hygroscopic
  • -60 Mesh Powder

Formula: Cs₂O₄S
MW: 361.87 g/mol
Boiling Pt: 1627
Melting Pt: 1010
Density: 4.243 g/mL
MDL Number: MFCD00010959
CAS Number: 10294-54-9
EINECS: 233-662-6
Merck Index: 13,02025

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR