2-Mercapto-5-methyl-1,3,4-thiadiazole ≥99%

Supplier: BeanTown Chemical

130590-250G 130590-50G
BT130590-250GEA 244 USD
BT130590-250G BT130590-50G
2-Mercapto-5-methyl-1,3,4-thiadiazole ≥99%
2-Mercapto-5-methyl-1,3,4-thiadiazole

CAS: 29490-19-5; EC No: 249-667-1; MDL No: MFCD00038349 Powder; Molecular Formula: C3H4N2S2; MW: 132.21 Melting Point: 188-189°


  • Powder

Formula: C₃H₄N₂S₂
Melting Pt: 188…189 °C
MDL Number: MFCD00038349
CAS Number: 29490-19-5
EINECS: 249-667-1

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR