Benzyl-2,2,2-trichloroacetimidate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Trichloroacetimidic Acid Benzyl Ester, Benzyl 2,2,2-trichloroacetimidate
130530-25G
130530-50G
130530-5G
BT130530-25GEA
95
USD
BT130530-25G
BT130530-50G
BT130530-5G
Benzyl-2,2,2-trichloroacetimidate ≥98%
Benzyl-2,2,2-trichloroacetimidate
CAS: 81927-55-1; MDL No: MFCD00000805
Liquid; Molecular Formula: C9H8Cl3NO; MW: 252.52
Boiling Point: 106-114°/0.5 mmHg; Flash point:
- Refractive index: 1.545
- Moisture Sensitive
- Liquid
Formula:
C₉H₈Cl₃NO MW: 252.52 g/mol Boiling Pt: 106…114 °C/0.5 mmHg Density: 1.359 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00000805 CAS Number: 81927-55-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...