Cyclohexanol ≥99%
Supplier: BeanTown Chemical
CAS: 108-93-0; EC No: 203-630-6; MDL No: MFCD00003855; RTECS: GV7875000
Liquid; Molecular Formula: C6H12O; MW: 100.16
Melting Point: 20-22°; Boiling Point: 160-161°; Flash point: 68°C (154°F)
Density (g/mL): 0.948; Refractive Index: 1.465
Hygroscopic
- RTECS: GV7875000
- Refractive index: 1.465
- Hygroscopic
- Liquid
Formula:
C₆H₁₂O Boiling Pt: 160…161 °C Melting Pt: 20…22 °C Density: 0.948 g/mL Flash Pt: 68 °C (154 °F) |
MDL Number:
MFCD00003855 CAS Number: 108-93-0 EINECS: 203-630-6 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...