Polyethylene glycol 400
Supplier: BeanTown Chemical
Synonyms:
PEG 400
130495-250G
130495-50G
BT130495-250GEA
64.29
USD
BT130495-250G
BT130495-50G
Polyethylene glycol 400
Polyethylene glycol 400
CAS: 25322-69-4; MDL No: MFCD01779680
Liquid; Linear Formula: H[OCH(CH3)CH2]nOH; MW ≈400
Density (g/mL): 1.005
- Liquid
Formula:
H(OCH₂CH₂)nOH MW: 400 g/mol Density: 1.005 g/mL |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...