Ammonium tetrathiotungstate

Supplier: BeanTown Chemical
Warning

Synonyms: Ammonium tetrathiotungstate(VI), Ammonium tungsten sulphide

130320-1G 130320-5G
BT130320-1GEA 29 USD
BT130320-1G BT130320-5G
Ammonium tetrathiotungstate
Ammonium tetrathiotungstate

CAS: 13862-78-7; MDL No: MFCD00061410 Powder/Chunks; Linear Formula: (NH4)2WS4; MW: 348.18 Hygroscopic


  • Hygroscopic
  • Powder/Chunks

Formula: H₈N₂S₄W
MW: 348.158 g/mol
MDL Number: MFCD00061410
CAS Number: 13862-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR