4-Bromo-2-methoxyphenol ≥98%
Supplier: BeanTown Chemical
130280-1G
130280-5G
BT130280-1GEA
23
USD
BT130280-1G
BT130280-5G
4-Bromo-2-methoxyphenol ≥98%
4-Bromo-2-methoxyphenol
CAS: 7368-78-7; MDL No: MFCD00051937
Solid; Molecular Formula: C7H7BrO2; MW: 203.03
Melting Point: 34-37°
- Solid
Formula:
C₇H₇BrO₂ Melting Pt: 34…37 °C |
MDL Number:
MFCD00051937 CAS Number: 7368-78-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...