2-Fluoro-4-methoxyphenylboronic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Fluoro-4-methoxybenzeneboronic acid

130275-1G 130275-5G
BT130275-1GEA 105 USD
BT130275-1G BT130275-5G
2-Fluoro-4-methoxyphenylboronic acid ≥98%
2-Fluoro-4-methoxyphenylboronic acid

CAS: 162101-31-7; MDL No: MFCD04039323 Solid; Molecular Formula: C7H8BFO3; MW: 169.95


  • Solid

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD04039323
CAS Number: 162101-31-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR