Calcium sulfate dihydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Plaster of Paris

130265-1KG 130265-5KG
BT130265-1KGEA 85.75 USD
BT130265-1KG BT130265-5KG
Calcium sulfate dihydrate ≥99%
Calcium sulphate dihydrate

CAS: 10101-41-4; EC No: 234-988-1; MDL No: MFCD00015981; RTECS: EW4150000 -325 Mesh Powder; Linear Formula: CaSO4·2H2O; MW: 172.17 Hygroscopic


  • RTECS: EW4150000
  • Hygroscopic
  • -325 Mesh Powder

Formula: CaSO₄·2H₂O
MW: 172.17 g/mol
MDL Number: MFCD00149625
CAS Number: 10101-41-4
EINECS: 234-988-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR