2-Octyn-1-ol ≥98%
Supplier: BeanTown Chemical
130255-10G
130255-50G
BT130255-10GEA
72
USD
BT130255-10G
BT130255-50G
2-Octyn-1-ol ≥98%
2-Octyn-1-ol
CAS: 20739-58-6; MDL No: MFCD00039542
Liquid; Linear Formula: CH3(CH2)4C≡CCH2OH; Molecular Formula: C8H14O; MW: 126.20
Boiling Point: 76-78°/2 mmHg; Flash point: 91°C (196°F)
Density (g/mL): 0.88; Refractive Index: 1.456
- Refractive index: 1.456
- Liquid
Formula:
C₈H₁₄O MW: 126.2 g/mol Boiling Pt: 76…78 °C/2 mmHg Density: 0.88 g/mL Flash Pt: 91 °C (196 °F) |
MDL Number:
MFCD00039542 CAS Number: 20739-58-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...