1,3-Bis(diphenylphosphino)propanenickel(II) chloride

Supplier: BeanTown Chemical

Synonyms: [1,3-Bis(diphenylphosphino)propane]nickel (II) chloride, Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II), 1,3-Bis(diphenylphosphino)propane nickel(II)chloride, Dichloro[1,3-bis(diphenylphosphino)propane]nickel(II), [1,3-Bis(diphenylphosphino)propane]dichloronickel(II), [1,3-Bis(diphenylphosphino)propane]nickel(II) dichloride

130225-1G 130225-5G
BT130225-1GEA 23 USD
BT130225-1G BT130225-5G
1,3-Bis(diphenylphosphino)propanenickel(II) chloride
1,3-Bis(diphenylphosphino)propanenickel(II) chloride

CAS: 15629-92-2; MDL No: MFCD00015318 Powder; Molecular Formula: C27H26Cl2NiP2; MW: 542.03 Melting Point: 213° (decomposes)


  • Powder

Formula: C₂₇H₂₆Cl₂NiP₂
Melting Pt: 213 °C (dec.)
MDL Number: MFCD00015318
CAS Number: 15629-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR