Cesium trifluoroacetate

Supplier: BeanTown Chemical
Warning

130210-10G 130210-50G
BT130210-10GEA 85 USD
BT130210-10G BT130210-50G
Cesium trifluoroacetate
Cesium trifluoroacetate

CAS: 21907-50-6; MDL No: MFCD00051304 Crystalline; Linear Formula: CF2COOCs; MW: 245.92


  • Crystalline

Formula: C₂CsF₃O₂
MW: 245.92
MDL Number: MFCD00051304
CAS Number: 21907-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR