N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine ≥94%
Supplier: BeanTown Chemical
Synonyms:
Benzyl(methoxymethyl)[(trimethylsilyl)methyl]amine
130200-1G
130200-5G
BT130200-1GEA
46
USD
BT130200-1G
BT130200-5G
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine ≥94%
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine
CAS: 93102-05-7; MDL No: MFCD00674005
Liquid; Linear Formula: C6H5CH2N(CH2OCH3)CH2Si(CH3)3; Molecular Formula: C13H23NOSi; MW: 237.42
Boiling Point: 76°/0.3 mmHg; Flash point: 66°C (150°F)
Density (g/mL): 0.928; Refractive Index: 1.492
Light Sensitive, Moisture Sensitive
- Refractive index: 1.492
- Light Sensitive, Moisture Sensitive
- Liquid
Formula:
C₁₃H₂₃NOSi Boiling Pt: 76 °C/0.3 mmHg Density: 0.928 g/mL Flash Pt: 66 °C (150 °F) |
MDL Number:
MFCD00674005 CAS Number: 93102-05-7 |
Specification Test Results
Assay | 94% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...