Tungsten zirconium oxide ≥99.5%
Supplier: BeanTown Chemical
130180-250G
130180-50G
BT130180-250GEA
1148
USD
BT130180-250G
BT130180-50G
Tungsten zirconium oxide ≥99.5%
Tungsten zirconium oxide
CAS: 16853-74-0; EC No: 240-876-3; MDL No: MFCD00168096
-200 Mesh Powder; Linear Formula: ZrW2O8; MW: 586.92
Density (g/mL): 5.1
- -200 Mesh Powder
Formula:
ZrW₂O₈ MW: 586.92 g/mol Density: 5.1 g/mL |
MDL Number:
MFCD00168096 CAS Number: 16853-74-0 EINECS: 240-876-3 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...