Diethyl maleate ≥97%

Supplier: BeanTown Chemical

Synonyms: Maleic acid diethyl ester, Diethyl (Z)-2-butenedioate

130160-1KG 130160-5KG
BT130160-1KGEA 97.27 USD
BT130160-1KG BT130160-5KG
Diethyl maleate ≥97%
Diethyl maleate

CAS: 141-05-9; EC No: 205-451-9; MDL No: MFCD00009191; RTECS: ON1225000 Liquid; Linear Formula: CH3CH2OCOCH=CHOCOCH2CH3; Molecular Formula: C8H12O4; MW: 172.18 Melting Point: -10°; Boiling Point: 225°; Flash point: 93°C (199°F) Density (g/mL): 1.064; Refractive Index: 1.441


  • RTECS: ON1225000
  • Refractive index: 1.441
  • Liquid

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 225 °C
Melting Pt: –10 °C
Density: 1.064 g/mL
Flash Pt: 93 °C (199 °F)
MDL Number: MFCD00009191
CAS Number: 141-05-9
EINECS: 205-451-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR