3,5-Dibromopyridine ≥98%

Supplier: BeanTown Chemical

130155-100G 130155-25G 130155-5G
BT130155-100GEA 958 USD
BT130155-100G BT130155-25G BT130155-5G
3,5-Dibromopyridine ≥98%
3,5-Dibromopyridine

CAS: 625-92-3; EC No: 210-916-4; MDL No: MFCD00014634 Solid; Molecular Formula: C5H3Br2N ; MW: 236.89 Melting Point: 110-115°


  • Solid

Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Melting Pt: 110…115 °C
MDL Number: MFCD00014634
CAS Number: 625-92-3
EINECS: 210-916-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR