1-Bromo-3-iodobenzene ≥98% stabilized

Supplier: BeanTown Chemical

130150-100G 130150-25G
BT130150-100GEA 313 USD
BT130150-100G BT130150-25G
1-Bromo-3-iodobenzene ≥98% stabilized
1-Bromo-3-iodobenzene

CAS: 591-18-4; EC No: 209-703-9; MDL No: MFCD00001043 Liquid; Molecular Formula: C6H4BrI; MW: 282.90 Melting Point: -9°; Boiling Point: 120°/18 mmHg; Flash point: 110°C (230°F) Density (g/mL): 2.219; Refractive Index: 1.660 Light Sensitive


  • Refractive index: 1.660
  • Light Sensitive
  • Liquid

Formula: C₆H₄BrI
Boiling Pt: 120 °C/18 mmHg
Melting Pt: –9 °C
Density: 2.219 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00001043
CAS Number: 591-18-4
EINECS: 209-703-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR