Digallium trioxide ≥99.999%

Supplier: BeanTown Chemical

130115-100G 130115-25G 130115-5G
BT130115-100GEA 739 USD
BT130115-100G BT130115-25G BT130115-5G
Digallium trioxide ≥99.999%
Digallium trioxide

CAS: 12024-21-4; EC No: 234-691-7; MDL No: MFCD00011020 -50 Mesh Powder; Molecular Formula: Ga2O3; MW: 187.44 Melting Point: 1900° Density (g/mL): 5.88


  • -50 Mesh Powder

Formula: Ga₂O₃
Melting Pt: 1900 °C
Density: 5.88 g/mL
MDL Number: MFCD00011020
CAS Number: 12024-21-4
EINECS: 234-691-7

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR