2-Fluoro-5-nitrobenzoic acid ≥98%

Supplier: BeanTown Chemical

130110-1G 130110-25G 130110-5G
BT130110-1GEA 27 USD
BT130110-1G BT130110-25G BT130110-5G
2-Fluoro-5-nitrobenzoic acid ≥98%
2-Fluoro-5-nitrobenzoic acid

CAS: 7304-32-7; MDL No: MFCD00134238 Solid; Molecular Formula: C7H4FNO4 ; MW: 185.11 Melting Point: 142-144°


  • Solid

Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Melting Pt: 142…144 °C
MDL Number: MFCD00134238
CAS Number: 7304-32-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR