2-(1-Adamantyl)-4-bromoanisole ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-(5-Bromo-2-methoxyphenyl)adamantane, 1-(5-Bromo-2-methoxy-phenyl)adamantane

130045-25G 130045-5G
BT130045-25GEA 561 USD
BT130045-25G BT130045-5G
2-(1-Adamantyl)-4-bromoanisole ≥98%
2-(1-Adamantyl)-4-bromoanisole

CAS: 104224-63-7; MDL No: MFCD03855308 Solid; Molecular Formula: C17H21BrO; MW: 321.25 Melting Point: 139-141°


  • Solid

Formula: C₁₇H₂₁BrO
MW: 321.2520 g/mol
Melting Pt: 139…141 °C
MDL Number: MFCD03855308
CAS Number: 104224-63-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR