2,4,6-Trimethylbenzaldehyde ≥98%
Supplier: BeanTown Chemical
Synonyms:
Mesitaldehyde
130035-25G
130035-5G
BT130035-25GEA
123
USD
BT130035-25G
BT130035-5G
2,4,6-Trimethylbenzaldehyde ≥98%
2,4,6-Trimethylbenzaldehyde
CAS: 487-68-3; EC No: 207-662-1; MDL No: MFCD00003341; RTECS: CU8500000
Liquid; Linear Formula: (CH3)3C6H2CHO; Molecular Formula: C10H12O; MW: 148.21
Melting Point: 10-12°; Boiling Point: 237°; Flash point: 106°C (222°F)
Density (g/mL): 1.005; Refractive Index: 1.553
Air Sensitive
- RTECS: CU8500000
- Refractive index: 1.553
- Air Sensitive
- Liquid
Formula:
C₁₀H₁₂O MW: 148.2 g/mol Boiling Pt: 237 °C Melting Pt: 10…12 °C Density: 1.005 g/mL Flash Pt: 106 °C (222 °F) |
MDL Number:
MFCD00003341 CAS Number: 487-68-3 EINECS: 207-662-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...