4-Iodoaniline ≥98%
Supplier: BeanTown Chemical
130025-250G
130025-50G
BT130025-250GEA
251
USD
BT130025-250G
BT130025-50G
4-Iodoaniline ≥98%
4-Iodoaniline
CAS: 540-37-4; EC No: 208-743-4; MDL No: MFCD00007848; RTECS: BY3850000
Crystalline; Molecular Formula: C6H6IN; MW: 219.03
Melting Point: 61-63°
Light Sensitive
- RTECS: BY3850000
- Light Sensitive
- Crystalline
Formula:
C₆H₆IN MW: 219.02 g/mol Melting Pt: 61…63 °C |
MDL Number:
MFCD00007848 CAS Number: 540-37-4 EINECS: 208-743-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...