D-(+)-Cellobiose ≥98%
Supplier: BeanTown Chemical
Synonyms:
β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose
130020-100G
130020-25G
130020-5G
BT130020-100GEA
206
USD
BT130020-100G
BT130020-25G
BT130020-5G
D-(+)-Cellobiose ≥98%
D(+)-Cellobiose
CAS: 528-50-7; EC No: 208-436-5; MDL No: MFCD00136034
Powder; Molecular Formula: C12H22O11; MW: 342.30
Melting Point: 239° (decomposes)
Optical Rotation: [α]20/D +34±1°, c = 10% in water
- Optical rotation: [α]20/D +34±1°, c = 10% in water
- Powder
Formula:
C₁₂H₂₂O₁₁ MW: 342.30 g/mol Melting Pt: 239 °C (dec.) |
MDL Number:
MFCD00136034 CAS Number: 528-50-7 EINECS: 208-436-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...