Sarcosine ≥98%

Supplier: BeanTown Chemical

Synonyms: H-Sar-OH, N-Methylglycine, Methylaminoacetic acid

130015-100G 130015-2.5KG 130015-500G
BT130015-100GEA 44.81 USD
BT130015-100G BT130015-2.5KG BT130015-500G
Sarcosine ≥98%
Sarcosine

CAS: 107-97-1; EC No: 203-538-6; MDL No: MFCD00004279; RTECS: VQ2897000 Crystalline; Linear Formula: CH3NHCH2COOH; Molecular Formula: C3H7NO2; MW: 89.09 Melting Point: 208-212° (decomposes) Hygroscopic


  • RTECS: VQ2897000
  • Hygroscopic
  • Crystalline

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Melting Pt: 208…212 °C (dec.)
MDL Number: MFCD00004279
CAS Number: 107-97-1
EINECS: 203-538-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR