S-(+)-Methyl-(2-chlorophenyl)[(2-(2-thienyl)amino]acetate hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: (S)-Methyl 2-(2-chlorophenyl)-2-((2-(thiophen-2-yl)ethyl)amino)acetate hydrochloride, 2-Chloro-N-[2-(2-thienyl)ethyl]-D-phenylglycine methyl ester hydrochloride

130010-1G 130010-250MG 130010-5G
BT130010-1GEA 176 USD
BT130010-1G BT130010-250MG BT130010-5G
S-(+)-Methyl-(2-chlorophenyl)[(2-(2-thienyl)amino]acetate hydrochloride ≥98%
S-(+)-Methyl-(2-chlorophenyl)[(2-(2-thienyl)amino]acetate hydrochloride

CAS: 141109-19-5; MDL No: MFCD08460122 Solid; Molecular Formula: C15H16ClNO2S·HCl; MW: 346.27


  • Solid

Formula: C₁₅H₁₇Cl₂NO₂S
MDL Number: MFCD08460122
CAS Number: 141109-19-5

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR