Diethylene glycol monomethyl ether ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-(2-Methoxyethoxy)ethanol, Diethylene glycol methyl ether

130005-2.5L 130005-500ML
BT130005-2.5LEA 69 USD
BT130005-2.5L BT130005-500ML
Diethylene glycol monomethyl ether ≥98%
Diethylene glycol monomethyl ether

CAS: 111-77-3; EC No: 203-906-6; MDL No: MFCD00002871; RTECS: KL6125000 Liquid; Linear Formula: CH3OCH2CH2ICH2CH2OH; Molecular Formula: C5H12O3; MW: 120.15 Melting Point: -70°; Boiling Point: 194°; Flash point: 87°C (189°F) Density (g/mL): 1.023; Refractive Index: 1.426 Hygroscopic


  • RTECS: KL6125000
  • Refractive index: 1.426
  • Hygroscopic
  • Liquid

Formula: C₅H₁₂O₃
MW: 120.15 g/mol
Boiling Pt: 194 °C
Melting Pt: –70 °C
Density: 1.023 g/mL
Flash Pt: 87 °C (189 °F)
MDL Number: MFCD00002871
CAS Number: 111-77-3
EINECS: 203-906-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR