Sorbitan monostearate
Supplier: BeanTown Chemical
Synonyms:
Sorbitan monooctadecanoate, (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol monooctadecanoate, Sorbitan stearate
130000-1KG
130000-250G
BT130000-1KGEA
78.78
USD
BT130000-1KG
BT130000-250G
Sorbitan monostearate
Sorbitan monostearate
CAS: 1338-41-6; EC No: 215-664-9; MDL No: MFCD00005366; RTECS: WG2933500
Powder; Molecular Formula: C24H46O6; MW: 430.62
Melting Point: 54-57°; Flash point:
- RTECS: WG2933500
- Powder
Formula:
C₂₄H₄₆O₆ MW: 430.63 g/mol Melting Pt: 54…57 °C Density: 1 g/mL Flash Pt: >149 °C (>300 °F) |
MDL Number:
MFCD00005366 CAS Number: 1338-41-6 EINECS: 215-664-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...