o-Anisic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Methoxybenzoic acid
129975-100G
129975-500G
BT129975-100GEA
41
USD
BT129975-100G
BT129975-500G
o-Anisic acid ≥98%
o-Anisic acid
CAS: 579-75-9; EC No: 209-447-8; MDL No: MFCD00002431; RTECS: BZ4385000
Powder; Molecular Formula: C8H8O3; MW: 152.15
Melting Point: 98-100°
- RTECS: BZ4385000
- Powder
Formula:
C₈H₈O₃ MW: 152.15 g/mol Melting Pt: 98…100 °C |
MDL Number:
MFCD00002431 CAS Number: 579-75-9 EINECS: 209-447-8 |
Specification Test Results
Assay | 98% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...