D-(+)-Raffinose pentahydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: D-(+)-Raffinose pentahydrate, Gossypose pentahydrate, (3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrate, Melitriose pentahydrate, O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose pentahydrate

129970-10G 129970-50G
BT129970-10GEA 29.34 USD
BT129970-10G BT129970-50G
D-(+)-Raffinose pentahydrate ≥99%
D-(+)-Raffinose pentahydrate

CAS: 17629-30-0; EC No: 208-146-9; MDL No: MFCD00071590; RTECS: LZ5851200 Crystalline/Powder; Molecular Formula: C18H32O16·5H2O; MW: 594.51 Hygroscopic


  • RTECS: LZ5851200
  • Hygroscopic
  • Crystalline/Powder

Formula: C₁₈H₃₂O₁₆·5H₂O
MW: 594.52 g/mol
MDL Number: MFCD00071590
CAS Number: 17629-30-0
EINECS: 208-146-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR