2-Bromo-5-(trifluoromethyl)pyridine ≥96%

Supplier: BeanTown Chemical

129925-1G 129925-250MG 129925-5G
BT129925-1GEA 85 USD
BT129925-1G BT129925-250MG BT129925-5G
2-Bromo-5-(trifluoromethyl)pyridine ≥96%
2-Bromo-5-(trifluoromethyl)pyridine

CAS: 50488-42-1; MDL No: MFCD00153086 Needles; Molecular Formula: C6H3BrF3N ; MW: 225.99 Melting Point: 44-48°


  • Needles

Formula: C₆H₃BrF₃N
MW: 226
Melting Pt: 44…48 °C
MDL Number: MFCD00153086
CAS Number: 50488-42-1

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR