4-Oxocyclohexanecarboxylic acid ≥98%
Supplier: BeanTown Chemical
129870-1G
129870-250MG
BT129870-1GEA
175
USD
BT129870-1G
BT129870-250MG
4-Oxocyclohexanecarboxylic acid ≥98%
4-Oxocyclohexanecarboxylic acid
CAS: 874-61-3; MDL No: MFCD00102035
Solid; Molecular Formula: C7H10O3; MW: 142.15
Melting Point: 67-71°
- Solid
Formula:
C₇H₁₀O₃ Melting Pt: 67…71 °C |
MDL Number:
MFCD00102035 CAS Number: 874-61-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...