Diethylenetriaminepentaacetic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: N-Carboxymethyliminobis(ethylenenitrilo)tetra(acetic acid), Kestranal® Aza Pa, DTPA, Titriplex® V, Idranal V, Diethylenetriaminepentaacetic acid, 2,2',2'',2'''-((((Carboxymethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetic acid, Diethylenetriamine-N,N,N',N'',N''-pentaacetic acid

129815-1KG 129815-250G 129815-50G
BT129815-1KGEA 107.5 USD
BT129815-1KG BT129815-250G BT129815-50G
Diethylenetriaminepentaacetic acid ≥98%
Diethylenetriaminepentaacetic acid

CAS: 67-43-6; EC No: 200-652-8; MDL No: MFCD00004289; RTECS: MB8205000 Powder; Molecular Formula: C14H23N3O10; MW: 393.35 Melting Point: 219-220°


  • RTECS: MB8205000
  • Powder

Formula: C₁₄H₂₃N₃O₁₀
MW: 393.35 g/mol
Melting Pt: 219…220 °C
MDL Number: MFCD00004289
CAS Number: 67-43-6
EINECS: 200-652-8
UN: 3077
ADR: 9 III
Merck Index: 13,07202

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR