Propylene carbonate ≥99%

Supplier: BeanTown Chemical

Synonyms: Propylene glycol carbonate, 4-Methyl-1,3-dioxolan-2-one, Carbonic acid propylene glycol ester

129800-1KG 129800-250G 129800-5KG
BT129800-1KGEA 52.93 USD
BT129800-1KG BT129800-250G BT129800-5KG
Propylene carbonate ≥99%
Propylene carbonate

CAS: 108-32-7; EC No: 203-572-1; MDL No: MFCD00005385 Liquid; Molecular Formula: C4H6O3; MW: 102.09 Melting Point: -55°; Boiling Point: 240-243°; Flash point: 132°C (270°F) Density (g/mL): 1.204; Refractive Index: 1.421 Hygroscopic, Moisture Sensitive


  • Refractive index: 1.421
  • Hygroscopic, Moisture Sensitive
  • Liquid

Formula: C₄H₆O₃
Boiling Pt: 240…243 °C
Melting Pt: –55 °C
Density: 1.204 g/mL
Flash Pt: 132 °C (270 °F)
MDL Number: MFCD00005385
CAS Number: 108-32-7
EINECS: 203-572-1

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR