(-)-Cinchonidine ≥99% may contain up to 5% quinine

Supplier: BeanTown Chemical

Synonyms: (8α,9R)-Cinchonan-9-ol

129795-100G 129795-25G
BT129795-100GEA 241 USD
BT129795-100G BT129795-25G
(-)-Cinchonidine ≥99% may contain up to 5% quinine
(-)-Cinchonidine

CAS: 485-71-2; EC No: 207-622-3; MDL No: MFCD00006783; RTECS: GD3500000 Powder; Molecular Formula: C19H22N2O; MW: 294.39 Melting Point: 204-206° Optical Rotation: [α]20/D -109°, c = 1.5 in ethanol Light Sensitive


  • RTECS: GD3500000
  • Optical rotation: [α]20/D -109°, c = 1.5 in ethanol
  • Light Sensitive
  • Powder

Formula: C₁₉H₂₂N₂O
MW: 294.4 g/mol
Melting Pt: 204…206 °C
MDL Number: MFCD00006783
CAS Number: 485-71-2
EINECS: 207-622-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR