2-Amino-5-pyridineboronic acid pinacol ester ≥97%
Supplier: BeanTown Chemical
Synonyms:
6-Aminopyridine-3-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 2-Aminopyridine-5-boronic acid pinacol ester
129780-1G
129780-5G
BT129780-1GEA
124
USD
BT129780-1G
BT129780-5G
2-Amino-5-pyridineboronic acid pinacol ester ≥97%
2-Amino-5-pyridineboronic acid pinacol ester
CAS: 827614-64-2; MDL No: MFCD05663837
Solid; Molecular Formula: C11H17BN2O2; MW: 220.08
Melting Point: 131-135°
- Solid
Formula:
C₁₁H₁₇BN₂O₂ Melting Pt: 131…135 °C |
MDL Number:
MFCD05663837 CAS Number: 827614-64-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...