3,4-Diethoxy-3-cyclobutene-1,2-dione ≥98%
Supplier: BeanTown Chemical
Synonyms:
Diethyl squarate, 3,4-Diethoxycyclobut-3-ene-1,2-dione
129770-1G
129770-25G
129770-5G
BT129770-1GEA
53
USD
BT129770-1G
BT129770-25G
BT129770-5G
3,4-Diethoxy-3-cyclobutene-1,2-dione ≥98%
3,4-Diethoxy-3-cyclobutene-1,2-dione
CAS: 5231-87-8; MDL No: MFCD00001333; RTECS: GU1772466
Liquid; Molecular Formula: C8H10O4; MW: 170.16
Boiling Point: 95°/0.1 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.15; Refractive Index: 1.509
- RTECS: GU1772466
- Refractive index: 1.509
- Liquid
Formula:
C₈H₁₀O₄ MW: 170.16 g/mol Boiling Pt: 95 °C/0.1 mmHg Density: 1.15 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00001333 CAS Number: 5231-87-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...