Potassium-1-carboxyvinyl hydrogenphosphate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Phosphoenolpyruvate potassium salt, Potassium 1-carboxyvinyl hydrogenphosphate, Phosphoenolpyruvic acid monopotassium salt, Phosphoenolpyruvic acid potassium
129745-100G
129745-25G
129745-5G
BT129745-100GEA
2821
USD
BT129745-100G
BT129745-25G
BT129745-5G
Potassium-1-carboxyvinyl hydrogenphosphate ≥99%
Potassium-1-carboxyvinyl hydrogenphosphate
CAS: 4265-07-0; EC No: 224-247-0; MDL No: MFCD00044476
Crystalline; Linear Formula: KOP(O)(OH)OC(=CH2)COOH; Molecular Formula: C3H4KO6P; MW: 206.14
Melting Point: 175° (decomposes)
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₃H₄KO₆P MW: 206.131 g/mol Melting Pt: 175 °C (dec.) |
MDL Number:
MFCD00044476 CAS Number: 4265-07-0 EINECS: 224-247-0 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...